missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ RAD51 Recombinant Protein

Código de producto. 16190012
Change view
Click to view available options
Cantidad:
10 μg
25 μg
Tamaño de la unidad:
10 microgramos
25 microgramos
Código de producto. Cantidad unitSize
16190012 10 μg 10 microgramos
16100022 25 μg 25 microgramos
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16190012

Marca: Abnova™ H00005888P01.10ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Human RAD51 full-length ORF recombinant protein with GST-tag at N-terminal

The protein encoded by this gene is a member of the RAD51 protein family. RAD51 family members are highly similar to bacterial RecA and Saccharomyces cerevisiae Rad51, and are known to be involved in the homologous recombination and repair of DNA. This protein can interact with the ssDNA-binding protein RPA and RAD52, and it is thought to play roles in homologous pairing and strand transfer of DNA. This protein is also found to interact with BRCA1 and BRCA2, which may be important for the cellular response to DNA damage. BRCA2 is shown to regulate both the intracellular localization and DNA-binding ability of this protein. Loss of these controls following BRCA2 inactivation may be a key event leading to genomic instability and tumorigenesis. Multiple transcript variants encoding different isoforms have been found for this gene

  • Theoretical MW (kDa): 52.36
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MAMQMQLEANADTSVEEESFGPQPISRLEQCGINANDVKKLEEAGFHTVEAVAYAPKKELINIKGISEAKADKILAVAERYGLSGSDVLDNVAYARAFNTDHQTQLLYQASAMMVESRYALLIVDSATALYRTDYSGRGELSARQMHLARFLRMLLRLADEFGVAVVITNQVVAQVDGAAMFAADPKKPIGGNIIAHASTTRLYLRKGRGETRICKIYDSPCLPEAEAMFAINADGVGDAKD

Best when used within three months from the date of receipt.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Especificaciones

Número de acceso AAH01459
Para utilizar con (aplicación) Antibody Production, Protein Array, ELISA, Western Blot
Formulación 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
ID de gen (Entrez) 5888
Peso molecular 52.36
Nombre RAD51 (Human) Recombinant Protein (P01)
Intervalo de pH 8
Método de preparación In vitro wheat germ expression system
Método de purificación Glutathione Sepharose 4 Fast Flow
Pruebas de control de calidad 12.5% SDS-PAGE Stained with Coomassie Blue.
Cantidad 10 μg
Fuente Wheat Germ (in vitro)
Inmunógeno MAMQMQLEANADTSVEEESFGPQPISRLEQCGINANDVKKLEEAGFHTVEAVAYAPKKELINIKGISEAKADKILAVAERYGLSGSDVLDNVAYARAFNTDHQTQLLYQASAMMVESRYALLIVDSATALYRTDYSGRGELSARQMHLARFLRMLLRLADEFGVAVVITNQVVAQVDGAAMFAADPKKPIGGNIIAHASTTRLYLRKGRGETRICKIYDSPCLPEAEAMFAINADGVGDAKD
Requisitos de almacenamiento Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Alias de gen BRCC5/HRAD51/HsRad51/HsT16930/RAD51A/RECA
Nombre común RAD51
Símbolo de gen RAD51
Reactividad cruzada Human
Especie Wheat Germ (in vitro)
Recombinante Recombinant
Etiqueta de proteína GST
Formulario Solution
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.