Learn More
Rabbit anti-Monkey Serum Proteins Secondary Antibody, Novus Biologicals™
Código de producto. 18613719 Tienda Bio Techne Productos
Change view
Click to view available options
:
1 mL; Unconjugated
Código de producto.
18613719 1 mL; Unconjugated
1 options
Código de producto. 18613719 Proveedor Novus Biologicals N.º de proveedor NBP269804
Rabbit Polyclonal Antibody
Serum Proteins Polyclonal antibody specifically detects Serum Proteins in Monkey samples. It is validated for ELISA, Immunoprecipitation, Immunoelectrophoresis
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Serum Proteins
Aplicaciones ELISA, Immunoprecipitation, Immunoelectrophoresis
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución ELISA, Immunoprecipitation 1:10 - 1:500, Immunoelectrophoresis
Formulación Whole antiserum
Especie del huésped Rabbit
Inmunógeno Pooled whole Rhesus monkey (Macaca mulatta) serum and partly purified serum fractions. Freund's complete adjuvant is used in the first step of the immunization procedure.
Método de purificación Whole Antiserum
Cantidad 1 mL
Estado normativo RUO
Primario o secundario Secondary
Reconstitución Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2).
Especies diana Monkey
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Lyophilized