missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
RAB9A Polyclonal antibody specifically detects RAB9A in Mouse samples. It is validated for Western Blot
Especificaciones
Especificaciones
| Antígeno | RAB9A |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Alexa Fluor 488 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | RAB9A, member RAS oncogene family, RAB9RAB9, member RAS oncogene family, ras-related protein Rab-9A |
| Especie del huésped | Rabbit |
| Inmunógeno | Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human RAB9A |
| Método de purificación | Antigen and protein A Affinity-purified |
| Cantidad | 0.1 mL |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?