missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RAB11FIP4 Monoclonal specifically detects RAB11FIP4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
| Antígeno | RAB11FIP4 |
| Aplicaciones | Western Blot, Immunohistochemistry |
| Clasificación | Monoclonal |
| Clon | OTI13B5 |
| Conjugado | Biotin |
| Dilución | Western Blot, Immunohistochemistry |
| Formulación | PBS with 0.05% Sodium Azide |
| Alias de gen | ARFO2, Arfophilin-2, FIP4-Rab11, FLJ00131, KIAA1821arfophilin-2, MGC11316, MGC126566, RAB11 family interacting protein 4 (class II), rab11 family-interacting protein 4, Rab11-FIP4 |
| Símbolos de los genes | RAB11FIP4 |
| Especie del huésped | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?