missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QPRT Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 25174-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
QPRT gene encodes a key enzyme in catabolism of quinolinate, an intermediate in the tryptophan-nicotinamide adenine dinucleotide pathway. Quinolinate acts as a most potent endogenous exitotoxin to neurons. Elevation of quinolinate levels in the brain has been linked to the pathogenesis of neurodegenerative disorders such as epilepsy, Alzheimer′s disease, and Huntington′s disease. Alternative splicing results in multiple transcript variants.Especificaciones
QPRT | |
Polyclonal | |
Unconjugated | |
QPRT | |
QAPRTase, QPRT, QPRTase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23475 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15274 | |
QPRT | |
QPRT Fusion Protein Ag17680 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |