missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Pyruvate Dehydrogenase E1-alpha subunit Polyclonal specifically detects Pyruvate Dehydrogenase E1-alpha subunit in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
| Antígeno | Pyruvate Dehydrogenase E1-alpha subunit |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Immunohistochemistry-Paraffin 1:500 - 1:1000 |
| Formulación | PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide |
| Alias de gen | EC 1.2.4.1, PDHA, PDHE1-A type I, PHE1APDHCE1A, pyruvate dehydrogenase (lipoamide) alpha 1, pyruvate dehydrogenase complex, E1-alpha polypeptide 1, pyruvate dehydrogenase E1 component subunit alpha, somatic form, mitochondrial |
| Símbolos de los genes | PDHA1 |
| Especie del huésped | Rabbit |
| Inmunógeno | This antibody was developed against a Recombinant Protein corresponding to amino acids:YCRSGKGPILMELQTYRYHGHSMSDPGVS |
| Vis mere |
For Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?