Learn More
PWP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca:  Proteintech 14767-1-AP-150UL
 Ver más versiones de este producto
Código de producto. 16853574
  • 410.00€ / 150 microlitros
Descripción
The protein encoded by this gene contains several WD-40 repeats and is found mostly in the nucleus. The expression and localization of this protein are cell cycle dependent. Expression of this gene is upregulated in pancreatic adenocarcinoma. Three transcript variants encoding different isoforms have been found for this gene.
Especificaciones
Especificaciones
PWP1
Polyclonal
Unconjugated
PWP1
IEF SSP 9502, PWP1, PWP1 homolog (S. cerevisiae)
Rabbit
Antigen Affinity Chromatography
RUO
103136, 11137, 362856
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13610, Q99LL5
Pwp1
PWP1 Fusion Protein Ag6533
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Sugerencias de productos
Sugerencias de productos
Videos
SDS
Documentos
Documentos
Certificados
Promociones
Promociones