missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
PUM2 Polyclonal antibody specifically detects PUM2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | PUM2 |
| Clasificación | Polyclonal |
| Dilución | Immunohistochemistry-Paraffin |
| Inmunógeno | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PUM2. |
| ID de gen (Entrez) | 23369 |
| Formulario | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?