missing translation for 'onlineSavingsMsg'
Learn More

PRPK Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Marca:  Novus Biologicals NBP1-69166

 Ver más versiones de este producto

Código de producto. 18265565

  • 466.00€ / 100 microlitros

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

PRPK Polyclonal specifically detects PRPK in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

PRPK
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
BUD32, C20orf64, chromosome 20 open reading frame 64, dJ101A2, dJ101A2.2, EC 2.7.11.1, Nori-2, Nori-2p, p53-related protein kinase, PRPK, TP53 regulating kinase, TP53-regulating kinase
Rabbit
28 kDa
100 μL
Protein Kinase
112858
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Western Blot, Immunocytochemistry, Immunofluorescence
0.5 mg/ml
Western Blot 1.0 ug/ml, Immunocytochemistry/Immunofluorescence
Q96S44
TP53RK
Synthetic peptides corresponding to TP53RK (TP53 regulating kinase) The peptide sequence was selected from the N terminal of TP53RK. Peptide sequence MAAARATTPADGEEPAPEAEALAAARERSSRFLSGLELVKQGAEARVFRG.
Affinity purified
RUO
Primary
Expected identity based on immunogen sequence: Human: 100%.
Human, Rat
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.