missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Progesterone Monoclonal antibody specifically detects Progesterone in Human samples. It is validated for Radioimmune Assays
Especificaciones
Especificaciones
| Antígeno | Progesterone |
| Aplicaciones | Radioimmune Assays (RIA) |
| Clasificación | Monoclonal |
| Clon | 11P14 |
| Concentración | 1 mg/mL |
| Conjugado | Unconjugated |
| Dilución | Radioimmunoassay 1:100 - 1:2000 |
| Formulación | PBS |
| Alias de gen | 4-Pregnen-3,20-dione |
| Especie del huésped | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?