missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prnd Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 51026-2-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This gene is found on chromosome 20, approximately 20 kbp downstream of the gene encoding cellular prion protein, to which it is biochemically and structurally similar. The protein encoded by this gene is a membrane glycosylphosphatidylinositol-anchored glycoprotein that is found predomitly in testis. Mutations in this gene may lead to neurological disorders.Specifications
Prnd | |
Polyclonal | |
Unconjugated | |
PRND | |
AI450264, doppel, Dpl, prion protein dublet, Prnd, PrPLP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23627, 26434 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9QUG3, Q9UKY0 | |
Prnd | |
Prnd Fusion Protein Ag0731 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |