Learn More
PP14/Glycodelin Mouse anti-Human, Clone: 16, Novus Biologicals™
Código de producto. 18582611 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mg
Tamaño de la unidad:
0.10 miligramo
18582611 0.1 mg 0.10 miligramo
1 options
Código de producto. 18582611 Proveedor Novus Biologicals N.º de proveedor NBP1051150.1mg
Mouse Monoclonal Antibody
PP14/Glycodelin Monoclonal specifically detects PP14/Glycodelin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno PP14/Glycodelin
Aplicaciones ELISA
Clasificación Monoclonal
Clon 16
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución ELISA 1:200, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulación 10mM Phosphate (pH 7.4) and 0.5M NaCl
Alias de gen alpha uterine protein, GdF, GdS, glycodelin, glycodelin-A, glycodelin-F, glycodelin-S, MGC138509, MGC142288, PEP, Placental protein 14, PP14 protein (placental protein 14), PP14GDPAEGGdA, pregnancy-associated endometrial a, Pregnancy-associated endometrial alpha-2 globulin, pregnancy-associated endometrial alpha-2-globulin, progestagen-associated endometrial protein (placental protein 14, progestagen-associated endometrial proteinPEG, Progesterone-associated endometrial protein
Especie del huésped Mouse
Inmunógeno Placental protein 14 (glycodelin A) purified from second trimester amniotic fluid
Método de purificación Protein A or G purified
Cantidad 0.1 mg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5047
Especies diana Human
Contenido y almacenamiento Store at 4°C. Do not freeze.
Formulario Purified
Isotype IgG1 κ