missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Solución estandarizada de hidróxido de potasio, 0,5N en metanol, Thermo Scientific Chemicals
Click to view available options
Cantidad:
1 L
4 L
20 L
Descripción
Potassium hydroxide, 0.5N standardized solution in methanol is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Especificaciones
Nombre del producto químico o material | Potassium hydroxide Standardized Solution |
Color | Incolora |
Número UN | UN3286 |
Concentración | 0.5N in methanol |
Información de solubilidad | Miscible with water. |
Nombre IUPAC | potassium hydroxide |
Formula Weight (peso de la fórmula) | 56.11 |
Olor | Like alcohol |
Cantidad | 4 L |
Forma física | Líquido |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'