missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Podocan Polyclonal antibody specifically detects Podocan in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | Podocan |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000 |
| Formulación | PBS (pH 7.3), 50% glycerol |
| Alias de gen | PCAN, PODOCAN, podocan proteoglycan, SLRR5AMGC24995 |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant fusion protein containing a sequence corresponding to amino acids 67-210 of human PODN (NP_714914.2). LGPVLAVRAPGFGRSGGHSLSPEENEFAEEEPVLVLSPEEPGPGPAAVSCPRDCACSQEGVVDCGGIDLREFPGDLPEHTNHLSLQNNQLEKIYPEELSRLHRLETLNLQNNRLTSRGLPEKAFEHLTNLNYLYLANNKLTLAP |
| Método de purificación | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?