Learn More
PMEL17/SILV Antibody (NKI-beteb), Novus Biologicals™
Código de producto. 18729193 Tienda Bio Techne Productos
Change view
Click to view available options
Tamaño de la unidad:
0.10 miligramo
20 microgramos
Cantidad:
20 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. unitSize Cantidad
18729193 0.10 miligramo -
18492122 20 microgramos 20 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18729193 Proveedor Novus Biologicals N.º de proveedor NBP229407
Mouse Monoclonal Antibody
PMEL17/SILV Monoclonal specifically detects PMEL17/SILV in Human, Equine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array, Flow (Intracellular).
TRUSTED_SUSTAINABILITY
Especificaciones
Especies diana Human, Equine
For Research Use Only