missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PM20D2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA556441
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: FVVPGIHPYF HIGSNALNHT EQYTEAAGSQ EAQFYTLRTA KALAMTALDV IFKPELLEGI REDFKLKLQE EQFVN Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.
PM20D2 belongs to the peptidase M20A family. The function of PM20D2 remains unknown.Especificaciones
PM20D2 | |
Polyclonal | |
Unconjugated | |
Pm20d2 | |
β-alanyl-lysine dipeptidase; Acy1l2; aminoacylase 1-like 2; aminoacylase 1-like protein 2; aminoacylase-1-like protein 2; bA63L7.3; Gm424; peptidase M20 domain containing 2; peptidase M20 domain-containing protein 2; Pm20d2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
135293 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IYS1 | |
Pm20d2 | |
Recombinant protein corresponding to Human PM20D2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only