missing translation for 'onlineSavingsMsg'
Learn More

PLK1 Antibody (CL8360) [CoraFluor™ 1], Novus Biologicals™

Código de producto. 30308059 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30308059 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30308059 Supplier Novus Biologicals Supplier No. NBP321166CL1

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

PLK1 Monoclonal antibody specifically detects PLK1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antígeno PLK1
Aplicaciones Western Blot
Clasificación Monoclonal
Clon CL8360
Conjugado CoraFluor 1
Formulación PBS
Alias de gen cell cycle regulated protein kinase, EC 2.7.11, EC 2.7.11.21, PLK, PLK-1, polo (Drosophia)-like kinase, polo like kinase, polo-like kinase 1polo-like kinase (Drosophila), Serine/threonine-protein kinase 13, serine/threonine-protein kinase PLK1, STPK13
Especie del huésped Mouse
Inmunógeno This antibody was developed using a synthetic peptide derived from P53350, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Phospho Specific, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 5347
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark. Do not freeze.
Formulario Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.