Learn More
PIWIL2 Antibody - BSA Free, Novus Biologicals™
Código de producto. 18142869 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.1 mg
Tamaño de la unidad:
0.03 miligramo
0.10 miligramo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18142869 0.1 mg 0.10 miligramo
18684486 0.025 mg 0.03 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18142869 Proveedor Novus Biologicals N.º de proveedor NBP241143
Rabbit Polyclonal Antibody
PIWIL2 Polyclonal specifically detects PIWIL2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno PIWIL2
Aplicaciones Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 5 ug/ml, Immunocytochemistry 2.5 ug/ml
Formulación PBS with 0.02% Sodium Azide
Alias de gen Cancer/testis antigen 80, CT80mili, FLJ10351, HILIMiwi like, MGC133049, Mili, PIWIL1L, piwil2-like protein, piwi-like 2 (Drosophila), piwi-like protein 2
Símbolos de los genes PIWIL2
Especie del huésped Rabbit
Inmunógeno Antibody was raised against a 17 amino acid synthetic peptide near the center of human PIWI-L2. The immunogen is located within amino acids 410 - 460 of PIWI-L2. Amino Acid Squence: TRYNNRTYRIDDVDWNK
Método de purificación Affinity Purified
Cantidad 0.1 mg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 55124
Especificidad de la prueba At least three isoforms of PIWI-L2 are known to exist; this antibody will only detect the two longest isoforms. PIWI-L2 antibody is predicted to not cross-react with other PIWI protein family members.
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG