missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PIP Monoclonal antibody specifically detects PIP in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antígeno | PIP |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | PIP/9076R |
| Conjugado | DyLight 350 |
| Formulación | 50 mM Sodium Borate |
| Alias de gen | GCDFP15prolactin-inducible protein, GCDFP-15SABP, GPIP4gp17, Gross cystic disease fluid protein 15, prolactin-induced proteinSecretory actin-binding protein |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant fragment (around aa 1-146) of human PIP protein (exact sequence is proprietary) |
| Método de purificación | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?