missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIM3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA567010
Detalles adicionales : Peso : 0.01000kg
Descripción
Immunogen sequence: FGTVYAGSRIA DGLPVAVKHV VKERVTEWGS LGGATVPLEV VLLR Highest antigen sequence identity to the following orthologs - mouse 96%, rat 61%.
PIM3 serine/threonine kinase or PIM-3 oncogene belongs to a family of protooncogenes. PIM3 is expressed in hepatocellular carcinoma development and plays a main role in the proliferation of human hepatoma cell lines. PIM3 also expressed in human pancreatic cancer cells and phosphorylates BAD to block Bad-mediated apoptosis in human pancreatic cancer cell lines. PIM3 is widely expressed, with the exception of no expression observed in colon, thymus, and small intestine.Especificaciones
PIM3 | |
Polyclonal | |
Unconjugated | |
PIM3 | |
BC026639; Kid1; KID-1/kinase induced by depolarization; Pim3; pim-3; pim-3 oncogene; Pim-3 proto-oncogene, serine/threonine kinase; proviral integration site 3; serine/threonine kinase Pim-3; serine/threonine-protein kinase pim-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
415116 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86V86 | |
PIM3 | |
Recombinant Human PIM3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only