missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PIK3C2G (PI3K-C2 gamma), GST Tag Recombinant Protein

Código de producto. p-7181647
Click to view available options
Cantidad:
100 μg
1 mg
20 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
20 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15970313

Marca: Invitrogen™ A31081

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. This PIK3C2G (PI3K-C2 gamma) recombinant human protein (468-1203) was expressed in insect cells. PIK3C2G (PI3K-C2 gamma) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service. Validated for use with the Adapta™ assay Available in multiple pack sizes for convenient use Also available in our SelectScreen Kinase Profiling Service Protein Form: Recombinant, Active, Catalytic The gene accession number for PIK3C2G (PI3K-C2 gamma) is AAI30278.1.

PIK3C2G lipid kinase belongs to the phosphoinositide 3-kinase (PI3K) family which play major roles in signaling pathways and are involved in cell proliferation, oncogenic transformation, cell survival, cell migration, and intracellular protein trafficking. PIK3C2G contains a lipid kinase catalytic domain as well as a C-terminal C2 domain, a characteristic of class II PI3-kinases that act as calcium-dependent phospholipid binding motifs which mediate translocation of proteins to membranes, and may also mediate protein-protein interactions. The N-terminal region of PIK3C2G and the C-terminal C2 domain are required for enzymatic activity. The PI 3-kinase C2gamma gene encodes a 1,050 amino acid polypeptide with 36% identity to human PI 3-kinase C2alpha. Research indicates that PI 3-kinase C2gamma can block the growth of human colon cancer cells.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso O75747
Concentración See Label
Para utilizar con (aplicación) Kinase Assay
Formulación 50mM tris HCl with 0.1mM PMSF, 0.25mM DTT, 25% glycerol, 0.1mM EDTA, 10mM glutathione, 150mM NaCl and no preservative; pH 7.5
ID de gen (Entrez) 5288
Peso molecular 110 kDa
Nombre Human PIK3C2G (PI3K-C2 gamma), GST Tag
Intervalo de pH 7.5
Método de purificación Purified
Cantidad 20 μg
Requisitos de almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Estado normativo RUO
Alias de gen C80387; phosphatidylinositol 3-kinase, C2 domain containing, gamma polypeptide; phosphatidylinositol 4-phosphate 3-kinase C2 domain-containing subunit gamma; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing gamma polypeptide; phosphatidylinositol-4-phosphate 3-kinase C2 domain-containing subunit gamma; phosphatidylinositol-4-phosphate 3-kinase catalytic subunit type 2 gamma; phosphatidylinositol-4-phosphate 3-kinase, catalytic subunit type 2 gamma; phosphoinositide 3-Kinase-C2-gamma; phosphoinositide-3-kinase, class 2, gamma polypeptide; PI 3-kinase C2gamma; PI3KC2gamma; PI3K-C2gamma; PI3K-C2-gamma; Pik3c2g; PTDINS-3-kinase C2 gamma; ptdIns-3-kinase C2 subunit gamma
Nombre común PIK3C2G
Símbolo de gen PIK3C2G
Familia Quinasa PI3 Kinase
Longitud de la proteína 468-1203
Tipo de producto Protein
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína GST-tag
Secuencia 468-1203
Contenido y almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Condiciones de envío Dry Ice
Sistema de expresión Insect cells
Familia de proteínas Kinases & Inhibitors
Forma de proteína Recombinant, Active, Catalytic
Formulario Liquid
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado