missing translation for 'onlineSavingsMsg'
Learn More

PHPT1 Antibody, Novus Biologicals™

Product Code. p-200060281 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Unit Size:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
18671256 100 μg 100 microlitros
18685725 50 μg 50 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18671256 Supplier Novus Biologicals Supplier No. NBP299999100ul

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PHPT1 Polyclonal antibody specifically detects PHPT1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno PHPT1
Aplicaciones Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunohistochemistry 1:500-1:2000, Immunoprecipitation 1-4 uL/mg of lysate, Immunohistochemistry-Paraffin
Formulación 0.2 um filtered solution in PBS
Alias de gen bA216L13.10, DKFZp564M173, EC 3.1.3,14 kDa phosphohistidine phosphatase, EC 3.1.3.-, HSPC141, phosphohistidine phosphatase 11700008C22Rik, phosphohistidine phosphatase 14kDa, PHP14CGI-202, Protein janus-A homolog, RP11-216L13.10, sex-regulated protein janus-a
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with purified, recombinant Human PHPT1 (Uniprot#: Q9NRX4-1; Ala2-Tyr125)
Método de purificación Antigen and protein A Affinity-purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Lipid and Metabolism
Primario o secundario Primary
ID de gen (Entrez) 29085
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.