missing translation for 'onlineSavingsMsg'
Learn More

Phosphoribosyl Pyrophosphate Amidotransferase Antibody (OTI1B11), Alexa Fluor™ 750, Novus Biologicals™

Código de producto. 18115117 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18115117 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18115117 Proveedor Novus Biologicals N.º de proveedor NBP273393AF750

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. View return policy

Mouse Monoclonal Antibody

Phosphoribosyl Pyrophosphate Amidotransferase Monoclonal specifically detects Phosphoribosyl Pyrophosphate Amidotransferase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Phosphoribosyl Pyrophosphate Amidotransferase
Aplicaciones Western Blot, Immunohistochemistry
Clasificación Monoclonal
Clon OTI1B11
Conjugado Alexa Fluor 750
Dilución Western Blot, Immunohistochemistry
Formulación 50 mM sodium borate with 0.05% sodium azide
Alias de gen ATASE, EC 2.4.2.14, glutamine phosphoribosylpyrophosphatate amidotransferase, Glutamine phosphoribosylpyrophosphate amidotransferase, glutamine PRPP amidotransferase, GPATamidophosphoribosyltransferase, phosphoribosyl pyrophosphate amidotransferase, PRAT
Símbolos de los genes PPAT
Especie del huésped Mouse
Inmunógeno Recombinant protein fragment corresponding to amino acids 42-278 of human Phosphoribosyl Pyrophosphate Amidotransferase (NP_002694) produced in E.coli.
Método de purificación Affinity Purified
Cantidad 0.1 mL
Disciplina de investigación Lipid and Metabolism
Primario o secundario Primary
ID de gen (Entrez) 5471.0
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.