Learn More
Phospholipase C like 1 Antibody, Novus Biologicals™
Código de producto. 18636297 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
18636297 50 μg 50 microlitros
18663047 100 μg 100 microlitros
2 options
Código de producto. 18636297
Proveedor: Novus Biologicals NBP30627550ul
Rabbit Polyclonal Antibody
Phospholipase C like 1 Polyclonal antibody specifically detects Phospholipase C like 1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Phospholipase C like 1
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Unconjugated
Dilución Immunohistochemistry-Paraffin 1:500-1:2000
Formulación PBS, pH7.0
Alias de gen but catalytically inactive protein, inactive phospholipase C-like protein 1, MGC126580, MGC138190, phospholipase C, epsilon, phospholipase C-deleted in lung carcinoma, phospholipase C-like 1, phospholipase C-related but catalytically inactive protein, PLCE, PLCL, PLC-L1, PRIP
Especie del huésped Rabbit
Inmunógeno Produced in rabbits immunized with E. coli-derived Human Phospholipase C like 1 fragment.
Método de purificación Antigen and protein A Affinity-purified
Cantidad 50 μg
Estado normativo RUO
Disciplina de investigación Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 5334
Especies diana Human
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG