missing translation for 'onlineSavingsMsg'
Learn More

Phospholipase B1 Antibody (OTI1D3), Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18105307 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
Product Code. Cantidad unitSize
18105307 0.1 mL 0.10 ml
1 options
This item is not returnable. View return policy

Product Code. 18105307

Brand: Novus Biologicals NBP273386AF750

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Phospholipase B1 Monoclonal specifically detects Phospholipase B1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Phospholipase B1
Aplicaciones Western Blot
Clasificación Monoclonal
Clon OTI1D3
Conjugado Alexa Fluor 750
Dilución Western Blot
Formulación 50 mM sodium borate with 0.05% sodium azide
Alias de gen FLJ30866, hPLB, phospholipase B1, PLBmembrane-associated
Símbolos de los genes PLB1
Especie del huésped Mouse
Inmunógeno Full length recombinant protein of human Phospholipase B1 (NP_694566) produced in HEK293T cells.
Método de purificación Affinity Purified
Cantidad 0.1 mL
Disciplina de investigación Lipid and Metabolism
Primario o secundario Primary
ID de gen (Entrez) 151056.0
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.