missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Phosphodiesterase 4D Monoclonal antibody specifically detects Phosphodiesterase 4D in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | Phosphodiesterase 4D |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | SR2286 |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000 |
| Formulación | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Alias de gen | cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E3), cAMP-specific 3'-5'-cyclic phosphodiesterase 4D, cAMP-specific phosphodiesterase PDE4D6, DKFZp686M11213, Drosophila), dunce-like phosphodiesterase E3, EC 3.1.4, FLJ97311, HSPDE4D, phosphodiesterase 4D, cAMP-specific, phosphodiesterase 4D, cAMP-specific (phosphodiesterase E3 dunce homolog |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthesized peptide derived from human phosphodiesterase 4D (Uniprot #: Q08499) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?