missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphodiesterase 4A/PDE4A Antibody (OTI1C8), PE/Cy5.5, Novus Biologicals™
Tienda Bio Techne ProductosDescripción
Phosphodiesterase 4A/PDE4A Monoclonal specifically detects Phosphodiesterase 4A/PDE4A in Human, Rat samples. It is validated for Flow Cytometry.
Especificaciones
Especificaciones
| Antígeno | Phosphodiesterase 4A/PDE4A |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | OTI1C8 |
| Conjugado | PE-Cyanine5.5 |
| Dilución | Flow Cytometry |
| Formulación | PBS with 0.05% Sodium Azide |
| Alias de gen | cAMP-specific 3′-5′-cyclic phosphodiesterase 4A, DPDE2cAMP-specific phosphodiesterase, EC 3.1.4, EC 3.1.4.17, PDE4, PDE46, phosphodiesterase 4A, cAMP-specific, phosphodiesterase 4A, cAMP-specific (dunce, phosphodiesterase 4A, cAMP-specific (dunce (Drosophila)-homologphosphodiesterase E2), phosphodiesterase E2 dunce homolog, Drosophila, phosphodiesterase isozyme 4 |
| Símbolos de los genes | PDE4A |
| Especie del huésped | Mouse |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?