missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGT Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 14327-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is expected to detect the endogenous PGT protein.
Prostaglandin transporter (PGT), also known as SLCO2A1, has been identified as a functional prostaglandin (PG) carrier that mediates either the release of newly-synthesized PGs by cells and/or the uptake of PGs prior to intracellular oxidation. Northern blot analysis revealed that PGT mRNA has broad tissue distribution, suggesting its role in the transport and/ or metabolic clearance of PG in diverse tissues (8787677).Especificaciones
| PGT | |
| Polyclonal | |
| Unconjugated | |
| SLCO2A1 | |
| OATP2A1, PGT, Prostaglandin transporter, SLC21A2, SLCO2A1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 24059, 6578 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q92959, Q9EPT5 | |
| SLCO2A1 | |
| PGT Fusion Protein Ag5594 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido