Learn More
Schmidlin™ PG Plus Hydrogen Generator
Descripción
- Increase efficiency and improve safety and performance
- On-demand supply 24/7
- Pressure: 11 barg (160 psig)
- Unique permeation membrane drying system
- USB connectivity
- Easy to install with complete 'plug and play' functionality
- Ideal for all GC detector applications
- Meets and exceeds requirements for the most demanding GC applications
- Superior hydrogen production with reliable long life cell
- Minimal maintenance; no desiccant cartridges to change
- PC monitoring for maintenance, diagnostics and remote connection
Especificaciones
Especificaciones
| Fondo (métrico) | 37 cm |
| Caudal | 100 mL/min |
| Altura (métrico) | 50.7 cm |
| Temperatura ambiente de funcionamiento máx. | 35°C |
| Presión de salida | 11 bar |
| Anchura (imperial) | 9 in. |
| Anchura (métrico) | 23 cm |
| Conexiones | Outlet: 1/8 in. |
| Requisitos eléctricos | 110/120 V at 60 Hz, 220/240 V at 50 Hz |
| Pureza del nitrógeno | >99.9996% |
| Mostrar más |