missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PEX5 Polyclonal antibody specifically detects PEX5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Specifications
Specifications
| Antígeno | PEX5 |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 0.04-0.4 μg/ml, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/ml, Immunohistochemistry-Paraffin 1:50 - 1:200, Knockdown Validated |
| Formulación | PBS (pH 7.2) and 40% Glycerol |
| N.º de referencia del gen | P50542 |
| Alias de gen | FLJ50634, FLJ50721, Peroxin-5, peroxisomal biogenesis factor 5, Peroxisomal C-terminal targeting signal import receptor, peroxisomal targeting signal 1 receptor, peroxisomal targeting signal import receptor, peroxisomal targeting signal receptor 1, Peroxisome receptor 1peroxin-5, PTS1 receptor, PTS1-BP, PTS1RFLJ51948, PXR1peroxisomal targeting signal 1 (SKL type) receptor |
| Especie del huésped | Rabbit |
| Inmunógeno | This antibody was developed against a recombinant protein corresponding to amino acids: RAQAEQWAAEFIQQQGTSDAWVDQFTRPVNTSALDMEFERAKSAIESDVDFWDKLQAELEEMAKRDAEAHPWLSDYDDLTSATYDKGYQFEEEN |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?