missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Peroxiredoxin 3 Monoclonal antibody specifically detects Peroxiredoxin 3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | Peroxiredoxin 3 |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | S07-3I3 |
| Concentración | 0.3 mg/mL |
| Conjugado | Unconjugated |
| Dilución | Western Blot, Immunohistochemistry 1:50, Immunohistochemistry-Paraffin |
| Formulación | 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Alias de gen | Antioxidant protein 1MGC104387, AOP-1MGC24293, AOP1PRO1748, EC 1.11.1.15, HBC189, MER5, peroxiredoxin 3, Peroxiredoxin III, peroxiredoxin-3, Protein MER5 homolog, prx-III, SP-22, thioredoxin-dependent peroxide reductase, mitochondrial |
| Especie del huésped | Rabbit |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?