missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Perilipin-3/TIP47 Polyclonal antibody specifically detects Perilipin-3/TIP47 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | Perilipin-3/TIP47 |
| Aplicaciones | Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | PE |
| Dilución | Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulación | PBS |
| Alias de gen | Cargo selection protein TIP47, M6PRBP1MGC2012, Mannose-6-phosphate receptor-binding protein 1, perilipin 3,47 kDa MPR-binding protein, perilipin-3, Placental protein 17,47 kDa mannose 6-phosphate receptor-binding protein, PP17MGC11117, tail-interacting protein, 47 kD, TIP47mannose-6-phosphate receptor binding protein 1 |
| Especie del huésped | Rabbit |
| Inmunógeno | Produced in rabbits immunized with E. coli-derived Human Perilipin-3/TIP47 fragment. |
| Método de purificación | Antigen and protein A Affinity-purified |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?