missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Pepsinogen A4/PGA4 Monoclonal antibody specifically detects Pepsinogen A4/PGA4 in Human samples. It is validated for ELISA
Especificaciones
Especificaciones
| Antígeno | Pepsinogen A4/PGA4 |
| Aplicaciones | ELISA |
| Clasificación | Monoclonal |
| Clon | 10 |
| Conjugado | Alexa Fluor 750 |
| Formulación | 50mM Sodium Borate |
| Especie del huésped | Mouse |
| Inmunógeno | This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Pepsinogen A4/PGA4 (Accession#: NP_001073276.1; Met1-Ala388). |
| Método de purificación | Protein A purified |
| Cantidad | 100 μg |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?