missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PeproGMP® Human IL-15 Recombinant Protein, PeproTech®

Código de producto. 30276215
Change view
Click to view available options
Cantidad:
100 μg
1 mg
50 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
50 microgramos
Código de producto. Cantidad unitSize
30276215 100 μg 100 microgramos
30275504 1 mg 1 miligramo
30275006 50 μg 50 microgramos
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30276215

Marca: Gibco™ GMP20015100UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Additional product information and specifications are provided on the Certificate of Analysis. All PeproGMP® products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter 1043 Ancillary Materials for Cell, Gene, and Tissue-Engineered Products. Product use statement: For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.

IL-15 (Interleukin 15 ) is a cytokine that regulates T and natural killer cell activation and proliferation. IL-15 and IL-2 share many biological activities as both have been found to bind common hematopoietin receptor subunits, and may compete for the same receptor, and thus negatively regulate each other's activity. The number of CD8+ memory cells is shown to be controlled by a balance between IL-15 and IL-2. IL-15 induces the activation of JAK kinases, as well as the phosphorylation and activation of transcription activators STAT3, STAT5, and STAT6. In mouse, studies suggest that IL-15 may increase the expression of apoptosis inhibitor Bcl-xL, possibly through the transcription activation activity of STAT6, and thus prevent apoptosis. IL-15 plays an important role in the growth and differentiation of T and B lymphocytes, natural killer cells, macrophages, and monocytes as well as activation of a number of important intracellular signaling molecules. As such, IL-15 could be essential for the immune responses, allograft rejection, and the pathogenesis of autoimmune diseases. Further, IL-15 is a widely expressed pro-inflammatory cytokine and has been shown to play a role in several inflammatory disorders, including rheumatoid arthritis, psoriasis and pulmonary inflammatory diseases. Emerging data suggest that IL-15 may serve as a good therapeutic target, as there appears to be a beneficial effect of IL-15 neutralization in models of psoriasis and diabetes.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P40933
Para utilizar con (aplicación) Control, Western Blot Control, Bioactivity
Formato Lyophilized
Formulación Protein with no preservative
ID de gen (Entrez) 3600
Peso molecular 12.9 kDa
Nombre PeproGMP™ Human IL-15
Método de purificación Purified
Cantidad 100 μg
Requisitos de almacenamiento -20°C or -80°C if preferred
Estado normativo GMP
Concentración de endotoxinas ≤0.1 EU/μg
Alias de gen AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15
Nombre común IL-15
Símbolo de gen IL15
Actividad biológica Determined by the proliferation of CTLL-2 cells. PeproGMP Recombinant Human IL-15 ha been tested against the WHO standard NIBSC 95/554.
Tipo de producto Protein
Conjugado Unconjugated
Recombinante Recombinant
Secuencia MNWVNVISDL KKIEDLIQSM HIDATLYTES DVHPSCKVTA MKCFLLELQV ISLESGDASI HDTVENLIIL ANNSLSSNGN VTESGCKECE ELEEKNIKEF LQSFVHIVQM FINTS
Contenido y almacenamiento -20°Cor to +80°C if preferred
Actividad Determined by the proliferation of CTLL-2 cells. PeproGMP Recombinant Human IL-15 has been tested against the WHO standard NIBSC 95/554. Find the International Unit (IU) value <a href='https://www.thermofisher.com/us/en/home/life-science/cell-culture/cell-culture-learning-center/recombinant-protein-information/international-units.html'>here</a>.
Sistema de expresión E. coli
Formulario Lyophilized
Línea de productos PeproGMP
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.