missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PeproGMP™ Human BMP-4 Recombinant Protein, PeproTech®

Código de producto. 30275492
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microgramos
50 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30275492

Marca: Gibco™ GMP12005ET50UG

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

Bone morphogenetic proteins (BMPs) constitute a subfamily within the TGF-beta superfamily of structurally related signaling proteins. PeproTech's E. coli-derived Human BMP-4 is a fully active homodimeric protein consisting of two 107 amino acid subunits, which correspond to amino acids 303-408 of the full-length BMP-4 precursor, as well as an N-terminal methionine. PeproTech Recombinant Human BMP-4 is a 24.2 kDa protein. PeproGMP BMP-4 can be used with PeproGMP VEGF-165 and PeproGMP SCF in CTS StemScale medium for pluripotent stem cell differentiation, specifically for mesoderm and hematopoetic progenitor cell induction. Reconstitute using 5mM HCl, pH 3.0 to a concentration of 0.1-1.0 mg/ml. Additional product information, specifications, handling and reconstitution information are provided on the Certificate of Analysis. All PeproGMP™ products are manufactured using animal-derived component free materials. Product Description/Intended Use: PeproTech GMP recombinant proteins are manufactured for use as ancillary materials by applying applicable principles of GMP and quality control requirements from USP (United States Pharmacopeia) Chapter <1043> Ancillary Materials for Cell, Gene, and Tissue-Engineered Products.

The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5' untranslated region of this gene has been described and three variants are described, all encoding an identical protein.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P12644
Para utilizar con (aplicación) Control, Western Blot Control, Bioactivity
Formato Lyophilized
Formulación Protein with no preservative
ID de gen (Entrez) 652
Peso molecular 24.2 kDa
Nombre PeproGMP™ Human BMP-4
Método de purificación Purified
Cantidad 50 μg
Requisitos de almacenamiento -20°C or -80°C if preferred
Estado normativo GMP
Concentración de endotoxinas ≤0.1 EU/μg
Alias de gen BMP; BMP2B; BMP-2B; Bmp2b1; Bmp2b-1; Bmp4; Bmp-4; BOMPR4A; Bone morphogenetic protein; Bone morphogenetic protein 2B; bone morphogenetic protein 4; bone morphogenetic protein 4 preproprotein; DVR4; Dvr-4; MCOPS6; OFC11; ZYME
Nombre común BMP4
Símbolo de gen BMP4
Actividad biológica Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells.
Tipo de producto Protein
Conjugado Unconjugated
Recombinante Recombinant
Secuencia MKKNKNCRRH SLYVDFSDVG WNDWIVAPPG YQAFYCHGDC PFPLADHLNS TNHAIVQTLV NSVNSSIPKA CCVPTELSAI SMLYLDEYDK VVLKNYQEMV VEGCGCR
Contenido y almacenamiento -20°Cor to +80°C if preferred
Actividad Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells.
Sistema de expresión E. coli
Formulario Lyophilized
Grado de pureza o calidad ≥ 98% by SDS-PAGE gel and HPLC analyses.
Mostrar más Mostrar menos
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado