missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PELP1 Antibody (4F1D1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NBP2-61778
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
PELP1 Monoclonal specifically detects PELP1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.
Especificaciones
| PELP1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000 | |
| HMX3, MNARmodulator of nongenomic activity of estrogen receptor, Modulator of non-genomic activity of estrogen receptor, P160, proline and glutamic acid rich nuclear protein, proline, glutamate and leucine rich protein 1, proline, glutamic acid and leucine rich protein 1, proline-, glutamic acid- and leucine-rich protein 1, proline-, glutamic acid-, leucine-rich protein 1, Transcription factor HMX3 | |
| Mouse | |
| 120 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 4F1D1 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| PELP1 | |
| Purified recombinant fragment of human PELP1 (AA: 1031-1180) expressed in E. Coli. | |
| Protein G purified | |
| Cellular Markers, Transcription Factors and Regulators | |
| 27043 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido