missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PDSS2 Mouse anti-Human, Clone: OTI1D12, liquid, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies TA503951
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.Especificaciones
PDSS2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q86YH6 | |
PDSS2 | |
Full length human recombit protein of human PDSS2 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI1D12 | |
Unconjugated | |
PDSS2 | |
C6orf210, COQ10D3, DLP1, bA59I9.3, hDLP1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
57107 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |