missing translation for 'onlineSavingsMsg'
Learn More

PDK4 Antibody, DyLight 550, Novus Biologicals™

Código de producto. 18269130 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
Código de producto. Cantidad unitSize
18269130 0.1 mL 0.10 ml
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18269130

Marca: Novus Biologicals NBP107047R

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

PDK4 Polyclonal specifically detects PDK4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno PDK4
Aplicaciones Western Blot, Immunocytochemistry, Immunofluorescence
Clasificación Polyclonal
Conjugado DyLight 550
Dilución Western Blot, Immunocytochemistry/Immunofluorescence
Formulación 50mM Sodium Borate with 0.05% Sodium Azide
N.º de referencia del gen Q16654
Alias de gen [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 4, mitochondrial, EC 2.7.11, EC 2.7.11.2, FLJ40832, Pyruvate dehydrogenase kinase isoform 4, pyruvate dehydrogenase kinase, isoenzyme 4, pyruvate dehydrogenase kinase, isozyme 4, pyruvate dehydrogenase, lipoamide, kinase isozyme 4, mitochondrial
Símbolos de los genes PDK4
Especie del huésped Rabbit
Inmunógeno Synthetic peptide made to an internal portion of the human PDK4 protein (within residues 100-150). [Swiss-Prot# Q16654]
Peso molecular del antígeno 49 kDa
Método de purificación Affinity Purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Lipid and Metabolism, Mitochondrial Markers, Protein Kinase, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 5166
Especificidad de la prueba Human and Mouse. Predicted to react with bovine based on 100% sequence homology. Immunogen sequence has 94% homology to rat.
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.