missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human PDGFRA (PDGFR alpha), GST Tag Recombinant Protein

Código de producto. p-7181618
Change view
Click to view available options
Cantidad:
10 μg
1 mg
Tamaño de la unidad:
1 miligramo
10 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
11543547 10 μg 10 microgramos
10260823 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 11543547 Proveedor Invitrogen™ N.º de proveedor PV3811

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Cytoplasmic.

PDGFRA is a cell surface receptor tyrosine kinase which binds members of the platelet-derived growth factor family. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. Studies in knockout mice, where homozygosity is lethal, indicate that the alpha form of the platelet-derived growth factor receptor is particularly important for kidney development since mice heterozygous for the receptor exhibit defective kidney phenotypes.
TRUSTED_SUSTAINABILITY

Especificaciones

Número de acceso P16234
Concentración See Label
Para utilizar con (aplicación) Kinase Assay
Formulación 50mM tris HCl with 2mM DTT, 0.5mM EDTA, 50% glycerol, 0.02% Triton X-100, 150mM NaCl and no preservative; pH 7.5
ID de gen (Entrez) 5156
Peso molecular 89.8 kDa
Nombre Human PDGFRA (PDGFR alpha), GST Tag
Intervalo de pH 7.5
Método de purificación Purified
Cantidad 10 μg
Requisitos de almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Estado normativo RUO
Alias de gen AI115593; alpha platelet-derived growth factor receptor; Alpha platelet-derived growth factor receptor precursor (PDGF-R-alpha); alpha-type platelet-derived growth factor receptor; APDGFR; CD140 antigen-like family member A; CD140A; CD140a antigen; GAS9; MGC74795; OTTHUMP00000218656; PDGF A-chain; PDGF alpha chain; PDGF Receptor alpha; PDGF subunit A; PDGF-1; PDGFACE; PDGFR2; Pdgfr-2; PDGFRA; pdgfr-a; PDGFRA/BCR fusion; PDGF-R-alpha; PDGFR-alpha; platelet derived growth factor receptor alpha; platelet derived growth factor receptor, alpha polypeptide; platelet-derived growth factor alpha receptor; Platelet-derived growth factor receptor 2; platelet-derived growth factor receptor alpha; platelet-derived growth factor receptor, alpha polypeptide; rearranged-in-hypereosinophilia-platelet derived growth factor receptor alpha fusion protein; RHEPDGFRA
Nombre común PDGFRA (CD140a)
Símbolo de gen PDGFRA
Familia Quinasa PDGFR Kinase Family
Longitud de la proteína 550-1089
Tipo de producto Protein
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína GST-tag
Secuencia 550-1089
Contenido y almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Entrada del ensayo Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical europium phospho-specific activity, Biochemical binding/kinetic
Grupo de cinasas Tyrosine Kinases
Condiciones de envío Dry Ice
Compatibilidad de sistemas LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
Entrada diana PDGFRA
Gen diana platelet-derived growth factor receptor, alpha polypeptide
Sistema de expresión Baculovirus
Familia de proteínas Kinases & Inhibitors
Forma de proteína Recombinant, Catalytic Domain, Cytoplasmic
Formulario Liquid
Subtipo de proteína Receptor Tyrosine Kinases
Categoría de investigación Signal Transduction
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.