missing translation for 'onlineSavingsMsg'
Learn More

PCNA Antibody (PCNA/8696R), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18582278 Shop All Bio Techne Products
Change view
Click to view available options
:
0.1 mL, Alexa Fluor 350
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18582278 0.1 mL, Alexa Fluor 350
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18582278 Supplier Novus Biologicals Supplier No. NBP320945AF350

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PCNA Monoclonal antibody specifically detects PCNA in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno PCNA
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon PCNA/8696R
Conjugado Alexa Fluor 350
Formulación 50 mM Sodium Borate
Alias de gen cyclin, DNA polymerase delta auxiliary protein, MGC8367, proliferating cell nuclear antigen
Especie del huésped Rabbit
Inmunógeno Recombinant full-length human PCNA protein
Método de purificación Protein A or G purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Autophagy, Base Excision Repair, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Polymerases, DNA Repair, Loading Controls, Phospho Specific, Stem Cell Markers
Primario o secundario Primary
ID de gen (Entrez) 5111
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.