missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax5/BSAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
195.00€
Especificaciones
| Antígeno | Pax5/BSAP |
|---|---|
| Concentración | 1 mg/mL |
| Dilución | Western Blot 1:500 - 1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunoprecipitation 2 - 5 μg, Immunohistochemistry-Paraffin 1:50 - 1:500 |
| Aplicaciones | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Clasificación | Polyclonal |
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|
18733204
|
Novus Biologicals
NBP2-29905-0.025mg |
0.025 mg | |||||||
Descripción
Pax5/BSAP Polyclonal specifically detects Pax5/BSAP in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Especificaciones
| Pax5/BSAP | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunoprecipitation 2 - 5 μg, Immunohistochemistry-Paraffin 1:50 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Q02548, Q02650 | |
| 5079 | |
| Purified internal fragment of human recombinant PAX5 expressed in E. coli | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Cancer, Transcription Factors and Regulators | |
| PBS, 1.0 mg/mL BSA and 30% Glycerol with 0.05% Sodium Azide | |
| B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5 | |
| PAX5 | |
| IgG | |
| Protein G purified |
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto