missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pax5/BSAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marca: Novus Biologicals NBP2-29905-0.025mg
Este artículo no se puede devolver.
Vea la política de devoluciones
Description
Pax5/BSAP Polyclonal specifically detects Pax5/BSAP in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Pax5/BSAP | |
| Polyclonal | |
| Unconjugated | |
| PBS, 1.0 mg/mL BSA and 30% Glycerol with 0.05% Sodium Azide | |
| B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:20 - 1:200, Immunoprecipitation 2 - 5 μg, Immunohistochemistry-Paraffin 1:50 - 1:500 | |
| Q02548, Q02650 | |
| PAX5 | |
| Purified internal fragment of human recombinant PAX5 expressed in E. coli | |
| 0.025 mg | |
| Cancer, Transcription Factors and Regulators | |
| 5079 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction