Learn More
Pax5/BSAP Antibody (CL14550) [Allophycocyanin], Novus Biologicals™
Código de producto. 30287384 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30287384 0.1 mL 0.10 ml
1 options
Código de producto. 30287384 Proveedor Novus Biologicals N.º de proveedor NBP326865APC
Mouse Monoclonal Antibody
Pax5/BSAP Monoclonal antibody specifically detects Pax5/BSAP in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno Pax5/BSAP
Aplicaciones Immunohistochemistry, Western Blot
Clasificación Monoclonal
Clon CL14550
Conjugado APC
Formulación PBS
Alias de gen B cell specific activator protein, B-cell-specific transcription factor, BSAPB-cell lineage specific activator, paired box 5, paired box gene 5 (B-cell lineage specific activator protein), paired box gene 5 (B-cell lineage specific activator), paired box homeotic gene 5, paired box protein Pax-5, transcription factor PAX 5
Especie del huésped Mouse
Inmunógeno This antibody was developed using a synthetic peptide derived from Q02548, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación B Cell Development and Differentiation Markers, Cancer, Transcription Factors and Regulators
Primario o secundario Primary
ID de gen (Entrez) 5079
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG2a