missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Parvalbumin alpha Antibody (PVALB/7601) [Alexa Fluor™ 700], Novus Biologicals™

Product Code. 30066500 Shop All Bio Techne Products
Change view
Click to view available options
Cantidad:
0.1 mL
Unit Size:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
30066500 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30066500 Supplier Novus Biologicals™ Supplier No. NBP327001AF700

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Parvalbumin alpha Monoclonal antibody specifically detects Parvalbumin alpha in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Parvalbumin alpha
Aplicaciones Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon PVALB/7601
Conjugado Alexa Fluor 700
Formulación 50 mM Sodium Borate
Alias de gen D22S749
Especie del huésped Mouse
Inmunógeno Recombinant fragment (around aa1-110) of human Parvalbumin alpha protein (exact sequence is proprietary)
Método de purificación Protein A or G purified
Cantidad 0.1 mL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 5816
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.