missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human PAK4, GST Tag Recombinant Protein
Click to view available options
Cantidad:
10 μg
Tamaño de la unidad:
10 microgramos
Descripción
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Catalytic
PAK4 is a serine/ threonine p21 activating kinase implicated in the reorganization of actin cytoskeleton and in the formation of filopodia. They are divided into two groups, the first of which include PAK1, 2 and 3, and can be activated by Cdc42/Rac binding. The second group includes PAK4, 5 and 6. These proteins are not activated by Cdc42/Rac binding. PAK4 was initially identified as a novel effector of Cdc42Hs. Co-expression of PAK4 and Cdc42Hs resulted in induction of filopodia and actin polymerization, showing that it is involved in cytoskeletal reorganization. Other experiments have shown PAK4 to be essential for embryonic viability and proper neuronal development. PAK4 has also been implicated in anchorage-independent growth of tumor cells and is required for activation of several cancer prosurvival pathways.
Especificaciones
Especificaciones
| Número de acceso | O96013 |
| Concentración | See Label |
| Para utilizar con (aplicación) | Kinase Assay |
| Formulación | 50 mM tris with 0.02% Triton X-100, 0.5 mM EDTA, 150 mM NaCl, 2 mM DTT, 50% glycerol and no preservative; pH 7.5 |
| ID de gen (Entrez) | 10298 |
| Peso molecular | 91.5 kDa |
| Nombre | Human PAK4, GST Tag |
| Cantidad | 10 μg |
| Estado normativo | For research use only. Not for use in diagnostic procedures |
| Alias de gen | 5730488L07Rik; AW555722; hypothetical protein LOC431769; im:5916170; Kiaa1142; mKIAA1142; p21 (CDKN1A)-activated kinase 4; p21 (RAC1) activated kinase 4; p21 protein (Cdc42/Rac)-activated kinase 4; p21(CDKN1A)-activated kinase 4; p21-activated kinase 4; Pak4; PAK-4; protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs; serine/threonine-protein kinase PAK 4; wu:fd12b01; zgc:92014 |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido