Learn More
Motic™ PA53 BIO Trinocular Research Upright Microscopes
Descripción
- Scalable Infinity-Corrected Optical System (CCIS™)
- Full power transmitted Köhler illumination
- Intelligent, heat-dissipating technology avoids overheating of microscope and sample
- Adjustable eyepieces and mutifunctional illumination control knob
- Environmentally friendly 'ECO' function provides automatic power-saving mode
- Homogeneous illumination with IL function
- Diopter Adjustment: ±4
Especificaciones
Especificaciones
| Eyepiece | WF 10X/23 mm |
| Estilo de cabeza | Trinocular |
| Tipo de iluminación | Transmitted Halogen |
| Potencia de aumento | 10X |
| Tipos de objetivos | Plano UC 4X, 10X, 40X, 100X |