missing translation for 'onlineSavingsMsg'
Learn More

p62/SQSTM1 Antibody - BSA Free, Novus Biologicals™

Código de producto. 18785663 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mL
0.1 mL
Tamaño de la unidad:
0.10 ml
25 microlitros
Código de producto. Cantidad unitSize
18785663 0.1 mL 0.10 ml
18797523 0.025 mL 25 microlitros
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18785663

Marca: Novus Biologicals NBP148320

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 75 publications

p62/SQSTM1 Polyclonal specifically detects p62/SQSTM1 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno p62/SQSTM1
Aplicaciones Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 1.0 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:4000, Simple Western 1:25, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:25-1:200, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating reported in scientific literature (PMID 35389045)
Formulación PBS with 0.02% Sodium Azide
N.º de referencia del gen Q13501, Q64337, O08623
Alias de gen A170, EBI3-associated protein of 60 kDa, EBI3-associated protein p60, EBIAP, ORCA, OSIL, oxidative stress induced like, p60PDB3, p62, p62B, Paget disease of bone 3, phosphotyrosine independent ligand for the Lck SH2 domain p62, Phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa, sequestosome 1, sequestosome-1, Ubiquitin-binding protein p62, ZIP3
Símbolos de los genes SQSTM1
Especie del huésped Rabbit
Inmunógeno Partial recombinant protein made to the C terminal of the human p62/SQSTM1 protein (within residues 300-440). [Swiss-Prot Q13501]
Método de purificación Affinity Purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Apoptosis, Autophagy, Cancer, mTOR Pathway, Neurodegeneration, Protein Turnover, Regulatory Immunology, Signal Transduction, Stem Cell Signaling Pathway, Ubiquitin Proteasome Pathway
Primario o secundario Primary
ID de gen (Entrez) 8878
Especificidad de la prueba Human, mouse and rat. Immunogen sequence has 98% homology to sheep.
Especies diana Human, Mouse, Rat, Chicken
Contenido y almacenamiento Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.