missing translation for 'onlineSavingsMsg'
Learn More

p21/CIP1/CDKN1A Antibody (CIP1/4377R), DyLight 594, Novus Biologicals™

Código de producto. 18528974 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
18528974 100 μg 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18528974

Marca: Novus Biologicals NBP308961DL594

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Monoclonal Antibody

p21/CIP1/CDKN1A Monoclonal antibody specifically detects p21/CIP1/CDKN1A in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antígeno p21/CIP1/CDKN1A
Aplicaciones Western Blot, Immunohistochemistry (Paraffin), Protein Array
Clasificación Monoclonal
Clon CIP1/4377R
Conjugado DyLight 594
Formulación 50mM Sodium Borate
Alias de gen CAP20cyclin-dependent kinase inhibitor 1, CDK-interacting protein 1, CDKN1melanoma differentiation associated protein 6, CIP1WAF1CDK-interaction protein 1, cyclin-dependent kinase inhibitor 1A (p21, Cip1), MDA6, MDA-6, Melanoma differentiation-associated protein 6, p21, p21CIP1, p21Cip1/Waf1, PIC1, SDI1DNA synthesis inhibitor, wild-type p53-activated fragment 1
Especie del huésped Rabbit
Inmunógeno Synthetic peptide corresponding to p21/CIP1/CDKN1A residues within aa1-100 of p21/CIP1/CDKN1A was used as an immunogen. (Uniprot: P38936 )
Método de purificación Protein A or G purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación Apoptosis, Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair
Primario o secundario Primary
ID de gen (Entrez) 1026
Especies diana Human
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.