missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p130Cas Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16815-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The p130Cas (Cas for Crk-associated substrate) is a common cellular target of phosphorylation signal via v-Crk and v-Src oncoproteins. p130Cas has a unique structure that contains a Src homology (SH)-3 domain followed by multiple YXXP motifs and a proline-rich regionp130Cas is implicated in a variety of biological processes including cell adhesion, cell migration, growth factor stimulation, cytokine receptor engagement and bacterial infection. Physiological functions of p130Cas include cardiovascular development, actin filament assembly and Src-induced cell transformation.Especificaciones
| p130Cas | |
| Polyclonal | |
| Unconjugated | |
| BCAR1 | |
| BCAR1, CAS, CAS1, CASS1, CRK associated substrate, CRKAS, p130Cas | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12927, 25414, 9564 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P56945, Q61140, Q63767 | |
| BCAR1 | |
| p130Cas Fusion Protein Ag10354 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido