missing translation for 'onlineSavingsMsg'
Learn More

p130Cas/BCAR1, p Tyr410 Antibody - BSA Free, Novus Biologicals™

Código de producto. 18611147 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
0.05 mg
0.1 mg
Tamaño de la unidad:
0.03 miligramo
0.05 miligramo
0.10 miligramo
Código de producto. Cantidad unitSize
18611147 0.1 mg 0.10 miligramo
18215753 0.05 mg 0.05 miligramo
18620387 0.025 mg 0.03 miligramo
3 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18611147

Marca: Novus Biologicals NBP254638

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

p130Cas/BCAR1 Polyclonal specifically detects p130Cas/BCAR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno p130Cas/BCAR1
Aplicaciones Western Blot, Immunohistochemistry
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100
Formulación Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, and 50% glycerol. with 0.02% sodium azide
Alias de gen breast cancer estrogen resistance 1, Cas scaffolding protein family member 1, CAS1, CASFLJ45059, CASS1CRKASbreast cancer estrogen resistance protein 1, Crkas, CRK-associated substrate, Crk-associated substrate p130Cas, FLJ12176, P130Cas
Símbolos de los genes BCAR1
Especie del huésped Rabbit
Inmunógeno p130 Cas (phospho Tyr410) antibody was raised against a peptide sequence around phosphorylation site of tyrosine 410 (G-V-Y (p)-A-V) derived from Human p130 Cas .
Método de purificación Affinity Purified
Cantidad 0.1 mg
Estado normativo RUO
Disciplina de investigación Breast Cancer, Cancer, MAP Kinase Signaling, Protein Kinase, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 9564
Especificidad de la prueba Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.